Gene Transcripts
- Gene Symbol
- Vps13c
- Entrez Gene ID
- 320528
- Gene Name
- vacuolar protein sorting 13C (yeast)
- Gene Aliases
- AI552542, AV026040, C230055H22Rik, W51662, chorein
- Chromosome Location
- Chr. 9: 67840331 - 67995640 on Build GRCm38
- UniGene ID
- Mm.241454
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_177184.3 |
NP_796158.2
|
37 |
4167 |
|
GenBank mRNA |
AB281675.1 |
- |
37 |
4167 |
|
View Full Product Details