Gene Transcripts

Gene Symbol
Gm14501
Entrez Gene ID
547154
Gene Name
predicted gene 14501
Gene Aliases
OTTMUSG00000016887
Chromosome Location
Chr. X: 9350599 - 9351137 on Build GRCm38
UniGene ID
Mm.327545
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085537.2 NP_001079006.1 1 232

Gene Symbol
1700012L04Rik
Entrez Gene ID
76383
Gene Name
RIKEN cDNA 1700012L04 gene
Gene Aliases
H2al1
Chromosome Location
Chr. X: 9283764 - 9284288 on Build GRCm38
UniGene ID
Mm.440761
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029588.3 NP_083864.1 1 230
GenBank mRNA AK005922.1 - 1 232

Gene Symbol
Gm14474
Entrez Gene ID
100042927
Gene Name
predicted gene 14474
Gene Aliases
OTTMUSG00000016779
Chromosome Location
Chr. X: 11302432 - 11302921 on Build GRCm38
UniGene ID
Mm.327561
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242947.1 NP_001229876.1 1 192

Gene Symbol
Gm14475
Entrez Gene ID
100042946
Gene Name
predicted gene 14475
Gene Aliases
ENSMUSG00000073279, OTTMUSG00000016780
Chromosome Location
Chr. X: 11324659 - 11324976 on Build GRCm38
UniGene ID
Mm.374311
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242954.1 NP_001229883.1 135

Gene Symbol
Gm14476
Entrez Gene ID
100042931
Gene Name
predicted gene 14476
Gene Aliases
ENSMUSG00000073284, OTTMUSG00000016781
Chromosome Location
Chr. X: 11308824 - 11309141 on Build GRCm38
UniGene ID
Mm.374311
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242950.1 NP_001229879.1 135

Gene Symbol
Gm14477
Entrez Gene ID
100042929
Gene Name
predicted gene 14477
Gene Aliases
OTTMUSG00000016783
Chromosome Location
Chr. X: 11305655 - 11305972 on Build GRCm38
UniGene ID
Mm.374311
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242949.1 NP_001229878.1 135

Gene Symbol
Gm14478
Entrez Gene ID
100042944
Gene Name
predicted gene 14478
Gene Aliases
OTTMUSG00000016784
Chromosome Location
Chr. X: 11321453 - 11321894 on Build GRCm38
UniGene ID
Mm.380300
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242953.1 NP_001229882.1 174

Gene Symbol
Gm14479
Entrez Gene ID
100042939
Gene Name
predicted gene 14479
Gene Aliases
OTTMUSG00000016785
Chromosome Location
Chr. X: 11315158 - 11315475 on Build GRCm38
UniGene ID
Mm.374311
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242951.1 NP_001229880.1 135

Gene Symbol
Gm14482
Entrez Gene ID
100042943
Gene Name
predicted gene 14482
Gene Aliases
OTTMUSG00000016788
Chromosome Location
Chr. X: 11318256 - 11318764 on Build GRCm38
UniGene ID
Mm.489576
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242952.1 NP_001229881.1 1 204

Gene Symbol
Gm14483
Entrez Gene ID
100042922
Gene Name
predicted gene 14483
Gene Aliases
OTTMUSG00000016789
Chromosome Location
Chr. X: 11299257 - 11299757 on Build GRCm38
UniGene ID
Mm.374311
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001111037.1 NP_001104507.1 1 199
GenBank mRNA BC087913.1 - 1 199

Gene Symbol
Gm14484
Entrez Gene ID
547160
Gene Name
predicted gene 14484
Gene Aliases
OTTMUSG00000016790
Chromosome Location
Chr. X: 11311934 - 11312427 on Build GRCm38
UniGene ID
Mm.350551
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025260.2 NP_001020431.1 1 192
GenBank mRNA BC049545.1 - 192

View Full Product Details