Gene Transcripts

Gene Symbol
Hist1h2ap
Entrez Gene ID
319171
Gene Name
histone cluster 1, H2ap
Gene Aliases
ENSMUSG00000060262, Gm11276, OTTMUSG00000000448
Chromosome Location
Chr. 13: 21833024 - 21833475 on Build GRCm38
UniGene ID
Mm.250564
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178185.2 NP_835492.1 1 116
GenBank mRNA AK145443.1 - 1 149
AK146846.1 - 1 147
BC062251.1 - 1 128 1224905
BC090402.1 - 1 128 5150365
BC110626.1 - 1 116 40051841
BC133661.1 - 1 116 40051853
BC147161.1 - 1 216 9056189
BC147162.1 - 1 216 9056203

Gene Symbol
Hist1h2ab
Entrez Gene ID
319172
Gene Name
histone cluster 1, H2ab
Gene Aliases
H2A.1, H2a-53
Chromosome Location
Chr. 13: 23751088 - 23751592 on Build GRCm38
UniGene ID
Mm.423402
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175660.3 NP_783591.2 1 161
GenBank mRNA BC117110.1 - 1 160

Gene Symbol
Hist1h2ad
Entrez Gene ID
319165
Gene Name
histone cluster 1, H2ad
Gene Aliases
Chromosome Location
Chr. 13: 23574381 - 23574915 on Build GRCm38
UniGene ID
Mm.390553
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178188.4 NP_835495.1 1 206
GenBank mRNA BC119295.1 - 1 206
BC119297.1 - 1 206 8734046

Gene Symbol
Hist1h2af
Entrez Gene ID
319173
Gene Name
histone cluster 1, H2af
Gene Aliases
H2a-221
Chromosome Location
Chr. 13: 23533911 - 23534378 on Build GRCm38
UniGene ID
Mm.377878
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175661.2 NP_783592.1 1 116
GenBank mRNA BC125011.1 - 1 116

Gene Symbol
Hist1h2ag
Entrez Gene ID
319167
Gene Name
histone cluster 1, H2ag
Gene Aliases
Chromosome Location
Chr. 13: 22042478 - 22042949 on Build GRCm38
UniGene ID
Mm.263165
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178186.3 NP_835493.1 1 138
GenBank mRNA BC065803.1 - 1 138

Gene Symbol
Hist1h2ah
Entrez Gene ID
319168
Gene Name
histone cluster 1, H2ah
Gene Aliases
Chromosome Location
Chr. 13: 22035122 - 22035643 on Build GRCm38
UniGene ID
Mm.250564
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175659.2 NP_783590.1 1 207
GenBank mRNA BC134370.1 - 1 116
BC139479.1 - 1 207 8862501
BC146008.1 - 1 207 40131311

Gene Symbol
Hist1h2ai
Entrez Gene ID
319191
Gene Name
histone cluster 1, H2ai
Gene Aliases
H2a-291A
Chromosome Location
Chr. 13: 21716412 - 21716859 on Build GRCm38
UniGene ID
Mm.14767
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178182.2 NP_835489.1 1 126
GenBank mRNA BC099406.1 - 126
BC116373.2 - 1 116 40100893
BC127164.1 - 1 116 40100895

Gene Symbol
Hist1h2an
Entrez Gene ID
319170
Gene Name
histone cluster 1, H2an
Gene Aliases
Chromosome Location
Chr. 13: 21786772 - 21787218 on Build GRCm38
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178184.2 NP_835491.1 1 116

Gene Symbol
Hist1h2ao
Entrez Gene ID
665433
Gene Name
histone cluster 1, H2ao
Gene Aliases
Chromosome Location
Chr. 13: 21810367 - 21810918 on Build GRCm38
UniGene ID
Mm.250564
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177544.2 NP_001171015.1 1 216

View Full Product Details