Gene Transcripts
- Gene Symbol
- MGRN1
- Entrez Gene ID
- 23295
- Gene Name
- mahogunin ring finger 1
- Gene Aliases
- RNF156
- Chromosome Location
- Chr. 16: 4616493 - 4690974 on Build GRCh38
- UniGene ID
- Hs.526494
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001142289.2 |
NP_001135761.2
|
5, 6 |
697 |
|
NM_001142290.2 |
NP_001135762.1
|
5, 6 |
697 |
|
NM_001142291.2 |
NP_001135763.2
|
5, 6 |
697 |
|
NM_015246.3 |
NP_056061.1
|
5, 6 |
697 |
|
NR_102267.1 |
|
5, 6 |
697 |
|
XM_005255217.1 |
XP_005255274.1
|
5, 6 |
697 |
|
XM_005255218.1 |
XP_005255275.1
|
5, 6 |
697 |
|
XM_005255219.3 |
XP_005255276.1
|
5, 6 |
697 |
|
XM_005255220.3 |
XP_005255277.1
|
5, 6 |
697 |
|
XM_005255221.3 |
XP_005255278.1
|
5, 6 |
697 |
|
XM_011522441.1 |
XP_011520743.1
|
5, 6 |
555 |
|
XM_017023091.1 |
XP_016878580.1
|
5, 6 |
697 |
|
XM_017023092.1 |
XP_016878581.1
|
5, 6 |
697 |
|
XM_017023093.1 |
XP_016878582.1
|
5, 6 |
555 |
|
GenBank mRNA |
AB011116.1 |
- |
5, 6 |
653 |
|
AK299052.1 |
- |
5, 6 |
679 |
|
BC050389.1 |
- |
5, 6 |
669 |
5755971 |
EF471397.2 |
- |
5, 6 |
560 |
|
EF471398.2 |
- |
5, 6 |
560 |
|
View Full Product Details