Gene Transcripts

Gene Symbol
RGD1565959
Entrez Gene ID
301230
Gene Name
RGD1565959
Gene Aliases
Chromosome Location
Chr. 9: 14883296 - 14896662 on Build Rnor_6.0
UniGene ID
Rn.92071
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106886.1 NP_001100356.1 2 284
XM_008766844.1 XP_008765066.1 2 193
XM_008766845.1 XP_008765067.1 2 193

View Full Product Details