Gene Transcripts
- Gene Symbol
- Hes5
- Entrez Gene ID
- 79225
- Gene Name
- hes family bHLH transcription factor 5
- Gene Aliases
- Chromosome Location
- Chr. 5: 172364421 - 172365188 on Build Rnor_6.0
- UniGene ID
- Rn.22422
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024383.1 |
NP_077359.1
|
1, 2 |
103 |
|
GenBank mRNA |
D12516.1 |
- |
1, 2 |
103 |
|
View Full Product Details