Gene Transcripts

Gene Symbol
Hiat1
Entrez Gene ID
100134827
Gene Name
hippocampus abundant transcript 1
Gene Aliases
Chromosome Location
Chr. 1: 69788609 - 69820986 on Build Rnor_6.0
UniGene ID
Rn.128804
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106467.3 NP_001099937.1 307
GenBank mRNA BC160912.1 - 296

Gene Symbol
NEWGENE_1305281
Entrez Gene ID
103689987
Gene Name
hippocampus abundant transcript 1
Gene Aliases
Hiat1
Chromosome Location
Chr. 2: 219661204 - 219697529 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006233223.2 XP_006233285.1 2, 3 439
XM_006233224.2 XP_006233286.1 2, 3 554
XM_008761473.1 XP_008759695.1 2, 3 310

View Full Product Details