Gene Transcripts
- Gene Symbol
- Olr1456
- Entrez Gene ID
- 405305
- Gene Name
- olfactory receptor 1456
- Gene Aliases
- Chromosome Location
- Chr. 10: 44730354 - 44731346 on Build Rnor_6.0
- UniGene ID
- Rn.142122
- Species
- Rattus norvegicus
- Gene Symbol
- Olr1452
- Entrez Gene ID
- 405048
- Gene Name
- olfactory receptor 1452
- Gene Aliases
- Chromosome Location
- Chr. 10: 44714212 - 44715201 on Build Rnor_6.0
- UniGene ID
- Rn.142126
- Species
- Rattus norvegicus
- Gene Symbol
- Olr1454
- Entrez Gene ID
- 287343
- Gene Name
- olfactory receptor 1454
- Gene Aliases
- Chromosome Location
- Chr. 11: 55483271 - 55483784 on Build Rnor_6.0
- UniGene ID
- Rn.142124
- Species
- Rattus norvegicus
View Full Product Details