Gene Transcripts

Gene Symbol
Cyp2b1
Entrez Gene ID
24300
Gene Name
cytochrome P450, family 2, subfamily b, polypeptide 1
Gene Aliases
Cyp2b-1, Cypbe
Chromosome Location
Chr. 7: 99142431 - 99183540 on Build Rnor_6.0
UniGene ID
Rn.228579
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134844.1 NP_001128316.1 1352
XM_008765491.1 XP_008763713.1 1395
XM_008765492.1 XP_008763714.1 1371
XM_008765493.1 XP_008763715.1 1757
GenBank mRNA BC166502.1 - 1352
J00719.1 - 1320
M37134.1 - 1320

Gene Symbol
Cyp2b2
Entrez Gene ID
361523
Gene Name
cytochrome P450, family 2, subfamily b, polypeptide 2
Gene Aliases
CYPIIB2, Cype
Chromosome Location
Chr. 1: 83103925 - 83119578 on Build Rnor_6.0
UniGene ID
Rn.228579
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001198676.1 NP_001185605.1 9 1359
GenBank mRNA K01626.1 - 2 183
M34452.1 - 8 1048

View Full Product Details