Gene Transcripts

Gene Symbol
NCSTN
Entrez Gene ID
23385
Gene Name
nicastrin
Gene Aliases
ATAG1874
Chromosome Location
Chr. 1: 160343056 - 160358952 on Build GRCh38
UniGene ID
Hs.517249
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290184.1 NP_001277113.1 4 512
NM_001290186.1 NP_001277115.1 3 373
NM_015331.2 NP_056146.1 3 373
XM_005245053.4 XP_005245110.1 3 590
XM_011509363.2 XP_011507665.1 3 590
GenBank mRNA AF240468.1 - 3 391
AK299142.1 - 3 260
AK310741.1 - 4 453
AK314764.1 - 3 280
AY359120.1 - 3 250
BC047621.1 - 4 469 4827966
D87442.1 - 3 247

View Full Product Details