Gene Transcripts

Gene Symbol
ZDHHC17
Entrez Gene ID
23390
Gene Name
zinc finger DHHC-type containing 17
Gene Aliases
HIP14, HIP3, HSPC294, HYPH
Chromosome Location
Chr. 12: 76764074 - 76853701 on Build GRCh38
UniGene ID
Hs.4014
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015336.2 NP_056151.2 9 1123
XM_005268750.4 XP_005268807.1 9 922
XM_011538083.1 XP_011536385.1 9 987
XM_017019091.1 XP_016874580.1 10 1073
XM_017019092.1 XP_016874581.1 9 936
XM_017019093.1 XP_016874582.1 9 885
XM_017019094.1 XP_016874583.1 8 803
XM_017019095.1 XP_016874584.1 5 515
GenBank mRNA AB023163.1 - 9 1067
AB024494.1 - 9 1613
AB097013.1 - 9 943
AB097035.1 - 9 943
AK292866.1 - 9 939
AK299089.1 - 9 945
BC050324.1 - 9 1082 5266097

View Full Product Details