Gene Transcripts

Gene Symbol
FBXO46
Entrez Gene ID
23403
Gene Name
F-box protein 46
Gene Aliases
20D7-FC4, FBXO34L, Fbx46
Chromosome Location
Chr. 19: 45710629 - 45730893 on Build GRCh38
UniGene ID
Hs.128702
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080469.1 NP_001073938.1 2 1708
GenBank mRNA BC021978.2 - 1 1096
BC046245.1 - 1 600
Y10936.1 - 1 256

View Full Product Details