Gene Transcripts

Gene Symbol
RIMBP2
Entrez Gene ID
23504
Gene Name
RIMS binding protein 2
Gene Aliases
PPP1R133, RBP2, RIM-BP2
Chromosome Location
Chr. 12: 130396133 - 130716361 on Build GRCh38
UniGene ID
Hs.657441
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015347.4 NP_056162.4 8 1187
XM_011538102.1 XP_011536404.1 9 1288
XM_011538103.2 XP_011536405.1 11 1750
XM_011538104.2 XP_011536406.1 11 1565
XM_011538105.2 XP_011536407.1 11 1739
XM_011538106.2 XP_011536408.1 11 1739
XM_011538107.2 XP_011536409.1 11 1746
XM_011538108.2 XP_011536410.1 11 1745
XM_017019105.1 XP_016874594.1 11 1763
XM_017019106.1 XP_016874595.1 11 1739
XM_017019107.1 XP_016874596.1 11 1561
XM_017019108.1 XP_016874597.1 11 1744
XM_017019109.1 XP_016874598.1 9 1288
XM_017019110.1 XP_016874599.1 11 1561
XM_017019111.1 XP_016874600.1 11 1561
XM_017019112.1 XP_016874601.1 11 1783
GenBank mRNA AB002316.1 - 8 1187
AY699979.1 - 5 878
BC007632.1 - 12 2074 3507574

View Full Product Details