Gene Transcripts

Gene Symbol
PCDHGA12
Entrez Gene ID
26025
Gene Name
protocadherin gamma subfamily A, 12
Gene Aliases
CDH21, FIB3, PCDH-GAMMA-A12
Chromosome Location
Chr. 5: 141430591 - 141512979 on Build GRCh38
UniGene ID
Hs.368160
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003735.2 NP_003726.1 1 1108
NM_032094.1 NP_115265.1 1 1108
GenBank mRNA AB011160.1 - 1 1081
AF152321.1 - 1 939
AF152506.1 - 1 939
AY358946.1 - 1 1309

View Full Product Details