Gene Transcripts

Gene Symbol
TSPAN17
Entrez Gene ID
26262
Gene Name
tetraspanin 17
Gene Aliases
FBX23, FBXO23, TM4SF17
Chromosome Location
Chr. 5: 176647387 - 176659058 on Build GRCh38
UniGene ID
Hs.532129
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001006616.2 NP_001006617.2 4 663
NM_012171.2 NP_036303.1 4 663
NM_130465.4 NP_569732.2 4 663
XM_005265876.3 XP_005265933.1 2 445
GenBank mRNA AK024427.1 - 4 553
AY037146.1 - 2 385
BC010405.2 - 4 485 3621871
BC067105.1 - 4 548 6646576

View Full Product Details