Gene Transcripts

Gene Symbol
FBXO3
Entrez Gene ID
26273
Gene Name
F-box protein 3
Gene Aliases
FBA, FBX3
Chromosome Location
Chr. 11: 33740937 - 33774531 on Build GRCh38
UniGene ID
Hs.406787
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012175.3 NP_036307.2 5 525
NM_033406.2 NP_208385.1 5 525
XM_011519980.1 XP_011518282.1 5 531
XM_011519981.2 XP_011518283.1 5 531
XM_011519982.1 XP_011518284.1 6 585
XM_017017533.1 XP_016873022.1 6 585
GenBank mRNA AB209039.1 - 4 3910
AF174595.1 - 5 493
AF176702.1 - 5 482
AK128454.1 - 5 503
AL136625.1 - 5 507
AL832826.1 - 4 1197
BC039291.1 - 5 525
BC046110.1 - 5 511 5549548
BG716664.1 - 5 528

View Full Product Details