Gene Transcripts

Gene Symbol
CD1D
Entrez Gene ID
912
Gene Name
CD1d molecule
Gene Aliases
CD1A, R3, R3G1
Chromosome Location
Chr. 1: 158178038 - 158186427 on Build GRCh38
UniGene ID
Hs.1799
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319145.1 NP_001306074.1 5 1130
NM_001766.3 NP_001757.1 6 1409
XM_005245583.1 XP_005245640.1 5 1088
XM_006711621.1 XP_006711684.1 5 970
XM_011510125.1 XP_011508427.1 6 1224
XM_011510127.1 XP_011508429.1 5 1052
GenBank mRNA AK225996.1 - 4 844
BC027926.1 - 6 1409 5229534
J04142.1 - 5 1074

View Full Product Details