Gene Transcripts
- Gene Symbol
- CPNE7
- Entrez Gene ID
- 27132
- Gene Name
- copine 7
- Gene Aliases
- Chromosome Location
- Chr. 16: 89575768 - 89597246 on Build GRCh38
- UniGene ID
- Hs.461775
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014427.4 |
NP_055242.1
|
13, 14 |
1472 |
|
NM_153636.2 |
NP_705900.1
|
11, 12 |
1247 |
|
XM_011523000.1 |
XP_011521302.1
|
11, 12 |
1242 |
|
XM_011523001.2 |
XP_011521303.1
|
11, 12 |
1007 |
|
XM_017023138.1 |
XP_016878627.1
|
13, 14 |
1436 |
|
XM_017023139.1 |
XP_016878628.1
|
13, 14 |
1447 |
|
XM_017023140.1 |
XP_016878629.1
|
11, 12 |
1222 |
|
XM_017023141.1 |
XP_016878630.1
|
10, 11 |
2329 |
|
GenBank mRNA |
AJ133798.1 |
- |
13, 14 |
1471 |
|
BC035334.1 |
- |
11, 12 |
1185 |
5191900 |
BC064577.1 |
- |
11, 12 |
1224 |
5727324 |
View Full Product Details