Gene Transcripts
- Gene Symbol
- GLS2
- Entrez Gene ID
- 27165
- Gene Name
- glutaminase 2
- Gene Aliases
- GA, GLS, LGA, hLGA
- Chromosome Location
- Chr. 12: 56470944 - 56488414 on Build GRCh38
- UniGene ID
- Hs.212606
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001280796.1 |
NP_001267725.1
|
7, 8 |
1138 |
|
NM_001280797.1 |
NP_001267726.1
|
6, 7 |
1008 |
|
NM_001280798.1 |
NP_001267727.1
|
4, 5 |
786 |
|
NM_013267.3 |
NP_037399.2
|
7, 8 |
1138 |
|
XM_005268797.1 |
XP_005268854.1
|
7, 8 |
765 |
|
XM_017019180.1 |
XP_016874669.1
|
6, 7 |
635 |
|
GenBank mRNA |
AF110330.1 |
- |
7, 8 |
893 |
|
AF223944.1 |
- |
7, 8 |
1065 |
|
AF279698.1 |
- |
4, 5 |
392 |
|
AK000467.1 |
- |
7, 8 |
885 |
|
AK294822.1 |
- |
7, 8 |
1083 |
|
AK303772.1 |
- |
7, 8 |
939 |
|
BC025661.1 |
- |
6, 7 |
723 |
|
DA060302.1 |
- |
4, 5 |
370 |
|
HM775422.1 |
- |
6, 7 |
757 |
|
HM775423.1 |
- |
5, 6 |
627 |
|
HM775424.1 |
- |
6, 7 |
757 |
|
View Full Product Details