Gene Transcripts

Gene Symbol
VENTXP1
Entrez Gene ID
139538
Gene Name
VENT homeobox pseudogene 1
Gene Aliases
CT18, NA88A, VENTX2P1
Chromosome Location
Chr. X: 26558337 - 26561052 on Build GRCh38
UniGene ID
Hs.447377
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_001559.2 1 166

Gene Symbol
VENTX
Entrez Gene ID
27287
Gene Name
VENT homeobox
Gene Aliases
HPX42B, NA88A, VENTX2
Chromosome Location
Chr. 10: 133237904 - 133241930 on Build GRCh38
UniGene ID
Hs.125231
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014468.3 NP_055283.1 3 1356
GenBank mRNA AF068006.1 - 3 1363

View Full Product Details