Gene Transcripts
- Gene Symbol
- UTP20
- Entrez Gene ID
- 27340
- Gene Name
- UTP20, small subunit processome component
- Gene Aliases
- 1A6/DRIM, DRIM
- Chromosome Location
- Chr. 12: 101280127 - 101386619 on Build GRCh38
- UniGene ID
- Hs.295732
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014503.2 |
NP_055318.2
|
41, 42 |
5545 |
|
GenBank mRNA |
AJ006778.1 |
- |
41, 42 |
5533 |
|
View Full Product Details