Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017829.5 | NP_060299.4 | 2 | 212 | |
NM_033070.2 | NP_149061.1 | 2 | 287 | ||
XM_005261236.3 | XP_005261293.1 | 2 | 213 | ||
XM_011546127.1 | XP_011544429.1 | 2 | 702 | ||
XM_017028781.1 | XP_016884270.1 | 2 | 702 | ||
XM_017028782.1 | XP_016884271.1 | 2 | 265 | ||
XM_017028783.1 | XP_016884272.1 | 2 | 702 | ||
XM_017028784.1 | XP_016884273.1 | 2 | 213 | ||
GenBank mRNA | AF273270.1 | - | 2 | 212 | |
AF273271.1 | - | 2 | 272 | ||
AK000461.1 | - | 2 | 285 | ||
AK315156.1 | - | 2 | 273 | ||
BC042540.1 | - | 2 | 271 | 4342582 |