Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001206924.1 | NP_001193853.1 | 3 | 432 | |
NM_001206925.1 | NP_001193854.1 | 3 | 718 | ||
NM_006889.4 | NP_008820.3 | 4 | 823 | ||
NM_175862.4 | NP_787058.4 | 4 | 768 | ||
NM_176892.1 | NP_795711.1 | 4 | 823 | ||
GenBank mRNA | AK291497.1 | - | 4 | 676 | |
AK294663.1 | - | 3 | 428 | ||
AK301237.1 | - | 3 | 714 | ||
AK313103.1 | - | 4 | 677 | ||
AK316203.1 | - | 3 | 714 | ||
BC040261.1 | - | 4 | 756 | 5173789 | |
CR541844.1 | - | 3 | 622 | ||
KU284848.1 | - | 3 | 622 | ||
L25259.1 | - | 4 | 746 | ||
U04343.1 | - | 4 | 769 |