Gene Transcripts

Gene Symbol
NTMT1
Entrez Gene ID
28989
Gene Name
N-terminal Xaa-Pro-Lys N-methyltransferase 1
Gene Aliases
AD-003, C9orf32, HOMT1A, METTL11A, NRMT, NTM1A
Chromosome Location
Chr. 9: 129608884 - 129636742 on Build GRCh38
UniGene ID
Hs.744027
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286796.1 NP_001273725.1 2, 3 504
NM_001286797.1 NP_001273726.1 2, 3 317
NM_001286798.1 NP_001273727.1 2, 3 378
NM_001286799.1 NP_001273728.1 2, 3 307
NM_001286800.1 NP_001273729.1 2, 3 352
NM_001286801.1 NP_001273730.1 2, 3 378
NM_014064.3 NP_054783.2 2, 3 352
GenBank mRNA AF110776.1 - 2, 3 295
AK290957.1 - 2, 3 349
AK292332.1 - 2, 3 354
AK298840.1 - 2, 3 342
AK313829.1 - 2, 3 342
BC001396.2 - 2, 3 331 3050940
BC033234.1 - 2, 3 306 5426447
BE392289.1 - 2, 3 309
BF316784.1 - 348
BG468453.1 - 2, 3 514
BI083998.1 - 2, 3 317
BM554138.1 - 2, 3 307
BQ923165.1 - 2, 3 331
CN406947.1 - 2, 3 365

View Full Product Details