Gene Transcripts

Gene Symbol
RBM15B
Entrez Gene ID
29890
Gene Name
RNA binding motif protein 15B
Gene Aliases
HUMAGCGB, OTT3
Chromosome Location
Chr. 3: 51391268 - 51397908 on Build GRCh38
UniGene ID
Hs.118738
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013286.4 NP_037418.3 1 2344
GenBank mRNA AL831838.1 - 1 1750
AY545557.1 - 1 318
BC001367.2 - 1 887
BC121184.2 - 2 225
BC139836.1 - 2 1555 40123447
L13434.1 - 1 2101

View Full Product Details