Gene Transcripts

Gene Symbol
CDR2L
Entrez Gene ID
30850
Gene Name
cerebellar degeneration related protein 2 like
Gene Aliases
HUMPPA
Chromosome Location
Chr. 17: 74987632 - 75005800 on Build GRCh38
UniGene ID
Hs.78358
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014603.2 NP_055418.2 3, 4 761
XM_006721852.2 XP_006721915.1 3, 4 746
GenBank mRNA AK294006.1 - 3, 4 761
BC047534.1 - 3, 4 761 5288296
L02867.1 - 3, 4 559

View Full Product Details