Gene Transcripts

Gene Symbol
EMC9
Entrez Gene ID
51016
Gene Name
ER membrane protein complex subunit 9
Gene Aliases
C14orf122, CGI-112, FAM158A
Chromosome Location
Chr. 14: 24138963 - 24142446 on Build GRCh38
UniGene ID
Hs.271614
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016049.3 NP_057133.2 5 594
XM_005267721.4 XP_005267778.1 4 1562
GenBank mRNA AF151870.1 - 5 589
AY368908.1 - 5 539
BC002491.2 - 5 594 3051105

View Full Product Details