Gene Transcripts

Gene Symbol
YBX2
Entrez Gene ID
51087
Gene Name
Y-box binding protein 2
Gene Aliases
CONTRIN, CSDA3, DBPC, MSY2
Chromosome Location
Chr. 17: 7288252 - 7294602 on Build GRCh38
UniGene ID
Hs.567494
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015982.3 NP_057066.2 9 1216
XM_017024713.1 XP_016880202.1 10 1306
GenBank mRNA AB208818.1 - 6 4197
AF096834.1 - 9 1214
AK092747.1 - 8 1833
BC033800.1 - 9 1179 5171696
BC047760.1 - 9 1180 5742121

View Full Product Details