Gene Transcripts

Gene Symbol
ZNF589
Entrez Gene ID
51385
Gene Name
zinc finger protein 589
Gene Aliases
SZF1
Chromosome Location
Chr. 3: 48241022 - 48275293 on Build GRCh38
UniGene ID
Hs.172602
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016089.2 NP_057173.2 4 919
XM_017006575.1 XP_016862064.1 4 1000
XM_017006576.1 XP_016862065.1 6 1149
XM_017006577.1 XP_016862066.1 4 2205
XM_017006578.1 XP_016862067.1 4 980
XM_017006579.1 XP_016862068.1 6 1105
GenBank mRNA AB209387.1 - 4 900
AF114816.1 - 6 1222
AF114817.1 - 6 1222
AL832831.1 - 4 904
AY258146.1 - 4 919
BC005859.2 - 4 878
BC006247.2 - 4 899
BC048798.1 - 4 896

View Full Product Details