Gene Transcripts

Gene Symbol
RAB23
Entrez Gene ID
51715
Gene Name
RAB23, member RAS oncogene family
Gene Aliases
HSPC137
Chromosome Location
Chr. 6: 57186992 - 57222314 on Build GRCh38
UniGene ID
Hs.555016
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278666.1 NP_001265595.1 5 645
NM_001278667.1 NP_001265596.1 5 817
NM_001278668.1 NP_001265597.1 5 644
NM_016277.4 NP_057361.3 5 1114
NM_183227.2 NP_899050.1 5 668
GenBank mRNA AB025427.1 - 4 460
AB034244.1 - 5 613
AF161486.1 - 5 610
AF498951.1 - 4 460
AY585189.1 - 4 460
BC015021.1 - 5 1080 3910708
CR749371.1 - 6 848

View Full Product Details