Gene Transcripts

Gene Symbol
NUP54
Entrez Gene ID
53371
Gene Name
nucleoporin 54
Gene Aliases
Chromosome Location
Chr. 4: 76114659 - 76148515 on Build GRCh38
UniGene ID
Hs.430435
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278603.1 NP_001265532.1 9 1252
NM_017426.3 NP_059122.2 10 1396
NR_103780.1 9 1169
NR_103781.1 8 1061
NR_103782.1 10 1166
XM_006714237.1 XP_006714300.1 11 1373
XM_011532033.2 XP_011530335.1 9 1231
XM_011532034.2 XP_011530336.1 9 1639
XM_017008294.1 XP_016863783.1 10 1783
XM_017008295.1 XP_016863784.1 9 1556
GenBank mRNA AF157322.1 - 10 1274
AK001517.1 - 10 1278
AK222752.1 - 10 1283
AK223352.1 - 10 1279
AK299442.1 - 9 1051
AK300036.1 - 9 1134
AK315160.1 - 10 1276
AL137650.1 - 3 395
BC012559.1 - 10 1293
BX538002.1 - 8 961

View Full Product Details