Gene Transcripts

Gene Symbol
ING3
Entrez Gene ID
54556
Gene Name
inhibitor of growth family member 3
Gene Aliases
Eaf4, ING2, MEAF4, p47ING3
Chromosome Location
Chr. 7: 120950755 - 120975657 on Build GRCh38
UniGene ID
Hs.489811
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019071.2 NP_061944.2 3 266
NM_198267.1 NP_938008.1 3 266
XM_017012369.1 XP_016867858.1 3 274
GenBank mRNA AF074968.1 - 3 243
AF161419.1 - 3 256
AF180298.1 - 3 233
AK000096.1 - 3 267
AK291905.1 - 3 244
AK293682.1 - 2 152
AL603623.1 - 3 243
AY007790.1 - 3 247
BC009776.1 - 3 221
BC009777.1 - 3 254
BC010851.1 - 3 213
BC062634.1 - 3 229
BC073865.1 - 3 234
BC093091.1 - 3 250
BC093689.1 - 3 226
BC101609.1 - 3 226
BC143316.1 - 3 226

View Full Product Details