Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001193307.1 | NP_001180236.1 | 2 | 2535 | |
NM_017654.3 | NP_060124.2 | 3 | 2636 | ||
GenBank mRNA | AB095925.2 | - | 3 | 2570 | |
AF445355.1 | - | 3 | 2636 | ||
AK122951.1 | - | 2 | 2473 | ||
AK125131.1 | - | 1 | 685 | ||
BC132773.1 | - | 2 | 2466 | 40146795 | |
BC132775.1 | - | 2 | 2466 | 40146797 | |
BC150249.1 | - | 3 | 2591 | 9007474 | |
BX647072.1 | - | 3 | 2592 |