Gene Transcripts

Gene Symbol
FBXL12
Entrez Gene ID
54850
Gene Name
F-box and leucine rich repeat protein 12
Gene Aliases
Fbl12
Chromosome Location
Chr. 19: 9810267 - 9820527 on Build GRCh38
UniGene ID
Hs.12439
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001316937.1 NP_001303866.1 2, 3 430
NM_001316940.1 NP_001303869.1 2, 3 352
NM_017703.2 NP_060173.1 2, 3 456
GenBank mRNA AK000195.1 - 2, 3 408
AK093760.1 - 2, 3 403
BC001586.2 - 2, 3 360 2966439
BP367273.1 - 2, 3 325
BX647073.1 - 1, 2 293
DB127388.1 - 2, 3 430

View Full Product Details