Gene Transcripts

Gene Symbol
SPDL1
Entrez Gene ID
54908
Gene Name
spindle apparatus coiled-coil protein 1
Gene Aliases
CCDC99
Chromosome Location
Chr. 5: 169583634 - 169604778 on Build GRCh38
UniGene ID
Hs.368710
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017785.4 NP_060255.3 11, 12 1955
XM_017009601.1 XP_016865090.1 11, 12 1786
XM_017009602.1 XP_016865091.1 10, 11 1591
GenBank mRNA AF269167.1 - 9, 10 1549
AK000371.1 - 10, 11 1682
AK074227.1 - 8, 9 1360
AK304623.1 - 10, 11 1619
AY517557.1 - 11, 12 1786
BC012568.1 - 11, 12 1821 4102558
BP295943.1 - 1, 2 85
CD653505.1 - 3, 4 604

View Full Product Details