Gene Transcripts

Gene Symbol
C19orf73
Entrez Gene ID
55150
Gene Name
chromosome 19 open reading frame 73
Gene Aliases
Chromosome Location
Chr. 19: 49118397 - 49119140 on Build GRCh38
UniGene ID
Hs.458310
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018111.2 NP_060581.2 1 683
GenBank mRNA AK001352.1 - 1 683
BC051004.1 - 1 688 6483373

View Full Product Details