Gene Transcripts

Gene Symbol
YY1AP1
Entrez Gene ID
55249
Gene Name
YY1 associated protein 1
Gene Aliases
HCCA1, HCCA2, YAP, YY1AP
Chromosome Location
Chr. 1: 155659442 - 155689032 on Build GRCh38
UniGene ID
Hs.584927
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001198899.1 NP_001185828.1 8 1854
NM_001198900.1 NP_001185829.1 10 1787
NM_001198901.1 NP_001185830.1 10 1677
NM_001198902.1 NP_001185831.1 10 1644
NM_001198903.1 NP_001185832.1 10 2164
NM_001198904.1 NP_001185833.1 10 2104
NM_001198905.1 NP_001185834.1 11 1768
NM_001198906.1 NP_001185835.1 11 1832
NM_018253.3 NP_060723.2 12 1909
NM_139118.2 NP_620829.1 11 1752
NM_139119.2 NP_620830.1 11 1825
NM_139121.2 NP_620832.1 11 1778
GenBank mRNA AF203474.1 - 10 1784
AF206328.1 - 10 1599
AF466401.1 - 10 1618
AK001737.1 - 7 1343
AK023976.1 - 11 1720
AK125590.1 - 4 1601
AK225883.1 - 7 1394
AK297562.1 - 10 1977
AK303386.1 - 8 1832
AK303944.1 - 6 1211
AK315049.1 - 11 1799
AY604179.1 - 10 1649
BC001655.2 - 12 1839 2961648
BC001843.2 - 12 1839 2961648
BC003500.1 - 11 1778
BC008766.2 - 10 1661 3051391
BC009358.2 - 7 1405
BC014906.2 - 11 1757 3922542
BC044887.1 - 10 1784 5276742
CR749789.1 - 11 1795

View Full Product Details