Gene Transcripts

Gene Symbol
DDX19A
Entrez Gene ID
55308
Gene Name
DEAD-box helicase 19A
Gene Aliases
DDX19-DDX19L, DDX19L
Chromosome Location
Chr. 16: 70346829 - 70373383 on Build GRCh38
UniGene ID
Hs.656037
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320522.1 NP_001307451.1 6 651
NM_001320525.1 NP_001307454.1 8 805
NM_001320526.1 NP_001307455.1 7 870
NM_001320527.1 NP_001307456.1 6 608
NM_018332.4 NP_060802.1 7 744
GenBank mRNA AB209401.1 - 1 1812
AK001988.1 - 7 702
AK223262.1 - 7 668
AK299504.1 - 4 334
AK299539.1 - 6 536
AK310604.1 - 6 532
AK310808.1 - 2 446
AL832901.1 - 3 228
AL832970.1 - 7 655
BC005162.2 - 7 652 3530064
BC006544.2 - 7 652 2964377
BC137496.1 - 7 630 9021496
BC137497.1 - 7 630 9021497
CR749227.1 - 7 817

View Full Product Details