Gene Transcripts

Gene Symbol
C5orf22
Entrez Gene ID
55322
Gene Name
chromosome 5 open reading frame 22
Gene Aliases
Chromosome Location
Chr. 5: 31532086 - 31555058 on Build GRCh38
UniGene ID
Hs.519246
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018356.2 NP_060826.2 2 333
NR_134298.1 2 333
XM_011514062.2 XP_011512364.1 2 318
XM_017009607.1 XP_016865096.1 2 322
XM_017009608.1 XP_016865097.1 2 333
XM_017009610.1 XP_016865099.1 2 322
XM_017009611.1 XP_016865100.1 2 322
XM_017009613.1 XP_016865102.1 2 513
XM_017009614.1 XP_016865103.1 2 315
GenBank mRNA AK002055.1 - 2 320
AK299156.1 - 2 310
BC021215.2 - 2 270 4302543
BC032845.1 - 2 316
DC404356.1 - 2 333

View Full Product Details