Gene Transcripts

Gene Symbol
SLC35C1
Entrez Gene ID
55343
Gene Name
solute carrier family 35 member C1
Gene Aliases
CDG2C, FUCT1
Chromosome Location
Chr. 11: 45804072 - 45813016 on Build GRCh38
UniGene ID
Hs.12211
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145265.1 NP_001138737.1 3 1132
NM_001145266.1 NP_001138738.1 3 1035
NM_018389.4 NP_060859.4 2 1406
XM_011520202.2 XP_011518504.1 2 293
GenBank mRNA AF323970.1 - 2 694
AF326199.1 - 2 1303
AK002182.1 - 3 808
AK027394.1 - 2 1383
AK027747.1 - 2 1347
AK074929.1 - 2 1407
AK091245.1 - 2 2090
AK225428.1 - 3 830
AK315473.1 - 2 1108
BC001427.2 - 2 711 3139599

View Full Product Details