Gene Transcripts

Gene Symbol
STYK1
Entrez Gene ID
55359
Gene Name
serine/threonine/tyrosine kinase 1
Gene Aliases
NOK, SuRTK106
Chromosome Location
Chr. 12: 10618923 - 10674318 on Build GRCh38
UniGene ID
Hs.24979
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018423.2 NP_060893.2 6, 7 1158
XM_005253417.2 XP_005253474.1 5, 6 1058
XM_011520736.1 XP_011519038.1 7, 8 961
XM_011520737.2 XP_011519039.1 6, 7 998
XM_011520738.2 XP_011519040.1 6, 7 1153
XM_017019579.1 XP_016875068.1 6, 7 941
GenBank mRNA AF251059.1 - 6, 7 1158
AK313906.1 - 6, 7 893
AL353940.1 - 6, 7 906
AY563054.1 - 4, 5 637
BC093822.1 - 4, 5 659 7939667
BC093824.1 - 4, 5 659 7939669
KP729000.1 - 4, 5 637

View Full Product Details