Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018425.3 | NP_060895.1 | 3 | 814 | |
GenBank mRNA | AJ303098.1 | - | 3 | 814 | |
AK309343.1 | - | 2 | 316 | ||
AL353952.1 | - | 1 | 95 | ||
BC003167.1 | - | 3 | 786 | 2905670 | |
BT007330.1 | - | 3 | 757 |