Gene Transcripts

Gene Symbol
AMBRA1
Entrez Gene ID
55626
Gene Name
autophagy and beclin 1 regulator 1
Gene Aliases
DCAF3, WDR94
Chromosome Location
Chr. 11: 46396412 - 46594069 on Build GRCh38
UniGene ID
Hs.654644
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267782.1 NP_001254711.1 10 2471
NM_001300731.1 NP_001287660.1 8 2462
NM_017749.3 NP_060219.2 9 2190
XM_005253009.3 XP_005253066.1 8 2458
XM_005253014.3 XP_005253071.1 9 2188
XM_006718259.2 XP_006718322.1 8 2339
XM_017018008.1 XP_016873497.1 8 2458
GenBank mRNA AB051523.1 - 8 2414
AK000301.1 - 10 2421
AK023197.1 - 2 713
AK302808.1 - 8 2520
AK304530.1 - 5 889
BC045609.1 - 9 2190 5296472

View Full Product Details