Gene Transcripts

Gene Symbol
UBAP2
Entrez Gene ID
55833
Gene Name
ubiquitin associated protein 2
Gene Aliases
UBAP-2
Chromosome Location
Chr. 9: 33921693 - 34048949 on Build GRCh38
UniGene ID
Hs.493739
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018449.3 NP_060919.3 11, 12 987
GenBank mRNA AK026088.1 - 3, 4 155
AK294952.1 - 11, 12 905
AK299183.1 - 5, 6 395
AK302335.1 - 9, 10 808
AK302754.1 - 10, 11 855
BC111762.1 - 5, 6 357

View Full Product Details