Gene Transcripts
- Gene Symbol
- CBWD1
- Entrez Gene ID
- 55871
- Gene Name
- COBW domain containing 1
- Gene Aliases
- COBP
- Chromosome Location
- Chr. 9: 121038 - 179083 on Build GRCh38
- UniGene ID
- Hs.531330
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001145355.1 |
NP_001138827.1
|
9, 10 |
862 |
|
NM_001145356.1 |
NP_001138828.1
|
8, 9 |
764 |
|
NM_018491.3 |
NP_060961.3
|
8, 9 |
764 |
|
XM_005251513.4 |
XP_005251570.1
|
7, 8 |
712 |
|
XM_005251517.4 |
XP_005251574.1
|
8, 9 |
764 |
|
XM_011517961.2 |
XP_011516263.1
|
9, 10 |
856 |
|
XM_011517962.2 |
XP_011516264.1
|
8, 9 |
764 |
|
XM_011517963.1 |
XP_011516265.1
|
5, 6 |
302 |
|
XM_017014925.1 |
XP_016870414.1
|
8, 9 |
764 |
|
GenBank mRNA |
AF212253.1 |
- |
8, 9 |
732 |
|
AF257330.2 |
- |
8, 9 |
722 |
|
AK290648.1 |
- |
9, 10 |
834 |
|
AK297292.1 |
- |
9, 10 |
803 |
|
AK315855.1 |
- |
8, 9 |
688 |
|
AY343911.1 |
- |
8, 9 |
721 |
|
BC005996.1 |
- |
8, 9 |
764 |
4069589 |
BC009573.1 |
- |
9, 10 |
815 |
4133536 |
BC013432.1 |
- |
8, 9 |
735 |
4133722 |
BC132759.1 |
- |
8, 9 |
698 |
40146781 |
- Gene Symbol
- CBWD3
- Entrez Gene ID
- 445571
- Gene Name
- COBW domain containing 3
- Gene Aliases
- bA561O23.1
- Chromosome Location
- Chr. 9: 68232437 - 68300035 on Build GRCh38
- UniGene ID
- Hs.645337
- Species
- Homo sapiens
- Gene Symbol
- CBWD5
- Entrez Gene ID
- 220869
- Gene Name
- COBW domain containing 5
- Gene Aliases
- DC36
- Chromosome Location
- Chr. 9: 65666374 - 65734041 on Build GRCh38
- UniGene ID
- Hs.355950
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286835.1 |
NP_001273764.1
|
9, 10 |
889 |
|
XM_005272748.1 |
XP_005272805.1
|
8, 9 |
761 |
|
XM_005272749.1 |
XP_005272806.1
|
8, 9 |
761 |
|
XM_005272750.1 |
XP_005272807.1
|
7, 8 |
701 |
|
XM_006716914.1 |
XP_006716977.1
|
8, 9 |
761 |
|
XM_017014456.1 |
XP_016869945.1
|
9, 10 |
692 |
|
XM_017014458.1 |
XP_016869947.1
|
8, 9 |
761 |
|
XM_017014459.1 |
XP_016869948.1
|
10, 11 |
940 |
|
GenBank mRNA |
AK097639.1 |
- |
7, 8 |
1077 |
|
AK292585.1 |
- |
9, 10 |
829 |
|
BC067803.1 |
- |
9, 10 |
827 |
5294979 |
BC141856.1 |
- |
9, 10 |
827 |
|
CD244615.1 |
- |
1, 2 |
68 |
|
View Full Product Details