Gene Transcripts

Gene Symbol
PCDHGC4
Entrez Gene ID
56098
Gene Name
protocadherin gamma subfamily C, 4
Gene Aliases
PCDH-GAMMA-C4
Chromosome Location
Chr. 5: 141485174 - 141512979 on Build GRCh38
UniGene ID
Hs.368160
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018928.2 NP_061751.1 1 941
NM_032406.1 NP_115782.1 1 941
GenBank mRNA AF152338.1 - 1 941
AF152525.1 - 1 941
BC042650.1 - 1 1033
BC101035.1 - 1 941 40007872
BC101036.1 - 1 938
BC101038.2 - 1 941 40007875

View Full Product Details