Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020165.3 | NP_064550.3 | 4 | 326 | |
GenBank mRNA | AB035274.2 | - | 4 | 361 | |
AF169796.1 | - | 4 | 306 | ||
AK023075.1 | - | 4 | 313 | ||
AK222771.1 | - | 4 | 304 | ||
AY004333.1 | - | 4 | 272 | ||
BC001302.1 | - | 4 | 293 | 3451960 |