Gene Transcripts

Gene Symbol
TTC7A
Entrez Gene ID
57217
Gene Name
tetratricopeptide repeat domain 7A
Gene Aliases
GIDID, MINAT, TTC7
Chromosome Location
Chr. 2: 46915931 - 47076136 on Build GRCh38
UniGene ID
Hs.370603
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001288951.1 NP_001275880.1 2 684
NM_001288953.1 NP_001275882.1 3 675
NM_001288955.1 NP_001275884.1 2 684
NM_020458.3 NP_065191.2 2 684
XM_005264439.3 XP_005264496.1 2 779
XM_011532998.2 XP_011531300.1 2 779
XM_011532999.2 XP_011531301.1 2 627
XM_017004524.1 XP_016860013.1 2 628
XM_017004525.1 XP_016860014.1 2 169
XM_017004526.1 XP_016860015.1 2 628
XM_017004527.1 XP_016860016.1 2 779
XM_017004528.1 XP_016860017.1 2 779
XM_017004529.1 XP_016860018.1 2 627
XM_017004530.1 XP_016860019.1 2 628
GenBank mRNA AB032966.2 - 1 74
AK056464.1 - 3 647
AL834383.1 - 2 684
BC111487.1 - 2 572 6041916
BC114365.2 - 2 315 40038218

View Full Product Details