Gene Transcripts

Gene Symbol
LYRM2
Entrez Gene ID
57226
Gene Name
LYR motif containing 2
Gene Aliases
DJ122O8.2
Chromosome Location
Chr. 6: 89632224 - 89638755 on Build GRCh38
UniGene ID
Hs.177275
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020466.4 NP_065199.1 3 262
NR_028493.1 3 421
NR_028494.1 3 404
NR_028495.1 3 272
GenBank mRNA AK024783.1 - 3 260
AK094994.1 - 3 330
AK311180.1 - 3 321
AK311948.1 - 3 247
AV710669.1 - 3 262
BC009782.1 - 3 256 4092710
BC104668.1 - 3 245 6717725
BP280644.1 - 3 262

View Full Product Details