Gene Transcripts

Gene Symbol
KLHL8
Entrez Gene ID
57563
Gene Name
kelch like family member 8
Gene Aliases
Chromosome Location
Chr. 4: 87160103 - 87220608 on Build GRCh38
UniGene ID
Hs.106601
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001292003.1 NP_001278932.1 4 1272
NM_001292006.1 NP_001278935.1 4 1001
NM_001292007.1 NP_001278936.1 3 680
NM_020803.4 NP_065854.3 4 1229
GenBank mRNA AB037799.2 - 3 912
AK128827.1 - 4 1272
BC041384.2 - 4 1179 5275946
BX538173.1 - 4 1146
BX640727.1 - 4 1139
BX640744.1 - 4 930
BX647454.1 - 4 1164
BX648367.1 - 2 197
CR749265.1 - 3 598

View Full Product Details